Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Mrpl32
Entrez Gene ID
291206
Gene Name
mitochondrial ribosomal protein L32
Gene Aliases
-
Chromosome Location
Chr.17: 53102534 - 53105400 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106116.1 NP_001099586.1 2 - 3 349 76

View Full Product Details