Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Slc2a8
Entrez Gene ID
85256
Gene Name
solute carrier family 2, (facilitated glucose transporter) member 8
Gene Aliases
Glut8
Chromosome Location
Chr.3: 12020227 - 12029918 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_053494.2 NP_445946.2 6 - 7 895 79
XM_008761779.1 XP_008760001.1 6 - 7 936 79
GenBank mRNA AJ245935.1 - 6 - 7 895 79

View Full Product Details