Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Nuak2
Entrez Gene ID
289419
Gene Name
NUAK family, SNF1-like kinase, 2
Gene Aliases
1200013b22rik, Snark, UV126
Chromosome Location
Chr.13: 49092306 - 49109055 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001007617.1 NP_001007618.1 1 - 2 315 95
GenBank mRNA BC081899.1 - 1 - 2 315 95

View Full Product Details