Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Otub1
Entrez Gene ID
293705
Gene Name
OTU deubiquitinase, ubiquitin aldehyde binding 1
Gene Aliases
-
Chromosome Location
Chr.1: 222451674 - 222459952 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106332.1 NP_001099802.1 4 - 5 361 69
GenBank mRNA BC166771.1 - 4 - 5 344 69

View Full Product Details