Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ctsw
Entrez Gene ID
293676
Gene Name
cathepsin W
Gene Aliases
-
Chromosome Location
Chr.1: 220844579 - 220847967 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001024242.1 NP_001019413.1 1 - 2 81 86
GenBank mRNA BC093401.1 - 1 - 2 81 86

View Full Product Details