Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Pwp1
Entrez Gene ID
362856
Gene Name
PWP1 homolog (S. cerevisiae)
Gene Aliases
RGD1310705
Chromosome Location
Chr.7: 23988776 - 24004774 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001191877.1 NP_001178806.1 12 - 13 1343 80

View Full Product Details