The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001108101.1 | NP_001101571.1 | 7 - 8 | 949 | 66 |
GenBank mRNA | BC166731.1 | - | 7 - 8 | 1024 | 66 |