The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001008344.1 | NP_001008345.1 | 10 - 11 | 1332 | 130 |
GenBank mRNA | BC085902.1 | - | 10 - 11 | 1332 | 130 |
FQ227237.1 | - | 10 - 11 | 1361 | 130 |