Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Amelx
Entrez Gene ID
29160
Gene Name
amelogenin, X-linked
Gene Aliases
Amel
Chromosome Location
Chr.X: 26439197 - 26450495 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001271073.1 NP_001258002.1 5 - 6 576 63
NM_001271074.1 NP_001258003.1 5 - 6 648 63
NM_001271075.1 NP_001258004.1 5 - 6 285 63
GenBank mRNA U51195.1 - 5 - 6 649 63
U60562.1 - 5 - 6 577 63
U60564.1 - 5 - 6 281 63

View Full Product Details