Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Slitrk4
Entrez Gene ID
302473
Gene Name
SLIT and NTRK-like family, member 4
Gene Aliases
-
Chromosome Location
Chr.X: 147582195 - 147589529 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106947.1 NP_001100417.1 1 - 2 247 56

View Full Product Details