Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Wnt1
Entrez Gene ID
24881
Gene Name
wingless-type MMTV integration site family, member 1
Gene Aliases
Int1
Chromosome Location
Chr.7: 140464999 - 140469046 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001105714.1 NP_001099184.1 3 - 4 953 68

View Full Product Details