The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001025686.1 | NP_001020857.1 | 4 - 5 | 595 | 79 |
GenBank mRNA | BC098725.1 | - | 4 - 5 | 595 | 79 |
FQ211503.1 | - | - | 620 | 79 |