Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Mtmr9
Entrez Gene ID
282584
Gene Name
myotubularin related protein 9
Gene Aliases
-
Chromosome Location
Chr.15: 46697262 - 46719296 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001005761.1 NP_001005761.1 6 - 7 1207 91
GenBank mRNA AF526269.1 - 6 - 7 88
BC083644.1 - 6 - 7 1207 91

View Full Product Details