Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tdh
Entrez Gene ID
290315
Gene Name
L-threonine dehydrogenase
Gene Aliases
-
Chromosome Location
Chr.15: 46667926 - 46681467 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106044.1 NP_001099514.1 2 - 3 258 80

View Full Product Details