Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Slc51a
Entrez Gene ID
303879
Gene Name
solute carrier family 51, alpha subunit
Gene Aliases
Osta, Ostalpha, RGD1311100
Chromosome Location
Chr.11: 71532787 - 71548506 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107087.1 NP_001100557.1 4 - 5 706 68
XM_008768725.1 XP_008766947.1 4 - 5 527 68

View Full Product Details