Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Lrrc58
Entrez Gene ID
303919
Gene Name
leucine rich repeat containing 58
Gene Aliases
RGD1307440
Chromosome Location
Chr.11: 65747235 - 65759558 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001195558.1 NP_001182487.1 1 - 2 532 142
GenBank mRNA CB547021.1 - 1 - 2 245 142

View Full Product Details