Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Itgb3bp
Entrez Gene ID
362548
Gene Name
integrin beta 3 binding protein (beta3-endonexin)
Gene Aliases
-
Chromosome Location
Chr.5: 118476410 - 118541928 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001013213.1 NP_001013231.1 3 - 4 517 59
XM_008763883.1 XP_008762105.1 4 - 5 259 59
XM_008763884.1 XP_008762106.1 3 - 4 213 59
GenBank mRNA BC086363.1 - 3 - 4 517 59

View Full Product Details