The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001017512.2 | NP_001017512.2 | 3 - 4 | 384 | 72 |
GenBank mRNA | BC079015.1 | - | 3 - 4 | 331 | 72 |
CB585199.1 | - | 3 - 4 | 384 | 72 |