Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Htatip2
Entrez Gene ID
292935
Gene Name
HIV-1 Tat interactive protein 2
Gene Aliases
-
Chromosome Location
Chr.1: 105094332 - 105109459 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106263.2 NP_001099733.1 4 - 5 514 86
XM_006229249.2 XP_006229311.1 5 - 6 665 86
XM_006229250.2 XP_006229312.1 5 - 6 569 86
XM_006229251.1 XP_006229313.1 5 - 6 580 86
GenBank mRNA BC158803.1 - 4 - 5 514 86

View Full Product Details