Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ugt2a3
Entrez Gene ID
289533
Gene Name
UDP glucuronosyltransferase 2 family, polypeptide A3
Gene Aliases
RGD1308444
Chromosome Location
Chr.14: 22251507 - 22267899 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001135869.1 NP_001129341.1 - 1301 80

View Full Product Details