The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_053935.1 | NP_446387.1 | 8 - 9 | 948 | 118 |
XM_006238359.2 | XP_006238421.1 | 8 - 9 | 2332 | 118 | |
GenBank mRNA | AF009603.1 | - | 5 - 6 | 550 | 118 |