Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Arl5b
Entrez Gene ID
364788
Gene Name
ADP-ribosylation factor-like 5B
Gene Aliases
Arl8
Chromosome Location
Chr.17: 82065910 - 82092693 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006254312.2 XP_006254374.1 4 - 5 582 81

View Full Product Details