The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001012173.1 | NP_001012173.1 | 7 - 8 | 782 | 55 |
XM_008760538.1 | XP_008758760.1 | 7 - 8 | 813 | 55 | |
GenBank mRNA | BC087683.1 | - | 7 - 8 | 782 | 55 |