Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Trim29
Entrez Gene ID
300656
Gene Name
tripartite motif-containing 29
Gene Aliases
-
Chromosome Location
Chr.8: 47674321 - 47699065 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106815.1 NP_001100285.1 4 - 5 1454 63

View Full Product Details