The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001127712.1 | NP_001121184.1 | 10 - 11 | 1470 | 104 |
GenBank mRNA | AY325176.1 | - | - | 1242 | 104 |
BC133725.1 | - | 10 - 11 | 1470 | 104 |