Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Siae
Entrez Gene ID
363045
Gene Name
sialic acid acetylesterase
Gene Aliases
RGD1310431
Chromosome Location
Chr.8: 40078269 - 40113514 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108759.1 NP_001102229.1 4 - 5 586 76
GenBank mRNA BC162050.1 - 4 - 5 557 76

View Full Product Details