Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ift27
Entrez Gene ID
300062
Gene Name
intraflagellar transport 27
Gene Aliases
Rabl4
Chromosome Location
Chr.7: 119393384 - 119409722 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001130495.1 NP_001123967.1 6 - 7 767 72
GenBank mRNA BC168150.1 - - 777 72

View Full Product Details