The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001011993.1 | NP_001011993.1 | 5 - 6 | 705 | 58 |
GenBank mRNA | BC087166.1 | - | 5 - 6 | 705 | 58 |