Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Invs
Entrez Gene ID
313228
Gene Name
inversin
Gene Aliases
RGD1563359
Chromosome Location
Chr.5: 64031131 - 64180830 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107932.1 NP_001101402.1 11 - 12 1965 66

View Full Product Details