The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_198740.2 | NP_942035.2 | 5 - 6 | 994 | 83 |
GenBank mRNA | BC091108.1 | - | 5 - 6 | 930 | 83 |
FQ226840.1 | - | 5 - 6 | 983 | 83 | |
FQ227160.1 | - | 5 - 6 | 931 | 83 | |
FQ233373.1 | - | 5 - 6 | 946 | 83 | |
Z49762.1 | - | 5 - 6 | 897 | 81 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_003751925.3 | XP_003751973.1 | 5 - 6 | 1031 | 83 |