Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Six6os1
Entrez Gene ID
500673
Gene Name
Six6 opposite strand transcript 1
Gene Aliases
RGD1559456
Chromosome Location
Chr.6: 95761797 - 95797304 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006240218.2 XP_006240280.1 2 - 3 91 132

View Full Product Details