Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Fshr
Entrez Gene ID
25449
Gene Name
follicle stimulating hormone receptor
Gene Aliases
-
Chromosome Location
Chr.6: 12795383 - 12997817 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_199237.1 NP_954707.1 8 - 9 733 119
XM_008764476.1 XP_008762698.1 6 - 7 393 119
GenBank mRNA L02842.1 - 8 - 9 733 119

View Full Product Details