Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pik3cg
Entrez Gene ID
298947
Gene Name
phosphatidylinositol-4,5-bisphosphate 3-kinase, catalytic subunit gamma
Gene Aliases
Pi3k
Chromosome Location
Chr.6: 51465696 - 51501192 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003750140.3 XP_003750188.1 5 - 6 2954 71
XM_006240003.2 XP_006240065.1 5 - 6 2527 71
XM_006240004.2 XP_006240066.1 5 - 6 2793 71
XM_006240005.2 XP_006240067.1 5 - 6 2603 71

View Full Product Details