The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001135749.1 | NP_001129221.1 | 2 - 3 | 222 | 63 |
GenBank mRNA | BC167077.1 | - | 2 - 3 | 222 | 63 |
BC168780.1 | - | 2 - 3 | 254 | 63 |