Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Edar
Entrez Gene ID
365581
Gene Name
ectodysplasin-A receptor
Gene Aliases
RGD1561714
Chromosome Location
Chr.20: 28179132 - 28263083 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001191899.1 NP_001178828.1 2 - 3 167 69
XM_006256381.2 XP_006256443.1 3 - 4 460 69
GenBank mRNA FQ211923.1 - 3 - 4 395 69

View Full Product Details