The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001014273.1 | NP_001014295.1 | 4 - 5 | 290 | 80 |
GenBank mRNA | BC083747.1 | - | 4 - 5 | 290 | 80 |