Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Alppl2
Entrez Gene ID
367308
Gene Name
alkaline phosphatase, placental-like 2
Gene Aliases
RGD1565100
Chromosome Location
Chr.9: 94174501 - 94181353 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_008767355.1 XP_008765577.1 10 - 11 2997 55

View Full Product Details