The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001012089.1 | NP_001012089.1 | 3 - 4 | 804 | 103 |
GenBank mRNA | BC088205.1 | - | 3 - 4 | 804 | 103 |