Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pan3
Entrez Gene ID
360760
Gene Name
PAN3 poly(A) specific ribonuclease subunit
Gene Aliases
-
Chromosome Location
Chr.12: 9214112 - 9331904 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006248842.2 XP_006248904.1 2 - 3 910 87
XM_006248844.2 XP_006248906.1 2 - 3 910 87
XM_006248845.2 XP_006248907.1 2 - 3 913 87
XM_346914.7 XP_346915.4 2 - 3 909 87

View Full Product Details