Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Nutm2f
Entrez Gene ID
306798
Gene Name
NUT family member 2F
Gene Aliases
Fam22f, RGD1566043
Chromosome Location
Chr.17: 15022698 - 15030483 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_008771552.1 XP_008769774.1 2 - 3 2665 110
XM_225188.7 XP_225188.5 3 - 4 1018 110

View Full Product Details