Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Slc6a16
Entrez Gene ID
292902
Gene Name
solute carrier family 6, member 16
Gene Aliases
RGD1561338
Chromosome Location
Chr.1: 101237539 - 101261671 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006229207.2 XP_006229269.1 5 - 6 1012 66

View Full Product Details