The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_001068704.5 | XP_001068704.3 | 5 - 6 | 899 | 71 |
XM_006249947.2 | XP_006250009.1 | 5 - 6 | 825 | 71 | |
XM_006249948.2 | XP_006250010.1 | 5 - 6 | 900 | 71 | |
XM_006249951.2 | XP_006250013.1 | 5 - 6 | 900 | 71 |