Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cfhr2
Entrez Gene ID
688887
Gene Name
complement factor H-related 2
Gene Aliases
-
Chromosome Location
Chr.13: 56641657 - 56713302 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001068704.5 XP_001068704.3 6 - 7 1093 117
XM_006249947.2 XP_006250009.1 6 - 7 1019 117
XM_006249948.2 XP_006250010.1 6 - 7 1094 117
XM_006249949.2 XP_006250011.1 5 - 6 916 117
XM_006249950.2 XP_006250012.1 4 - 5 733 117
XM_006249951.2 XP_006250013.1 6 - 7 1094 117
GenBank mRNA FQ211198.1 - 5 - 6 896 117

View Full Product Details