Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Zpld1
Entrez Gene ID
363768
Gene Name
zona pellucida-like domain containing 1
Gene Aliases
RGD1308138
Chromosome Location
Chr.11: 47924970 - 47973359 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006248257.1 XP_006248319.1 2 - 3 330 67

View Full Product Details