The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001105726.1 | NP_001099196.1 | 3 - 4 | 1040 | 150 |
GenBank mRNA | FQ219112.1 | - | 3 - 4 | 680 | 150 |