Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LOC500183
Entrez Gene ID
500183
Gene Name
similar to NGF-binding Ig light chain
Gene Aliases
D4Mit8, Igk@
Chromosome Location
-
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA BC098746.1 - - 204 161

View Full Product Details