Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
St3gal4
Entrez Gene ID
363040
Gene Name
ST3 beta-galactoside alpha-2,3-sialyltransferase 4
Gene Aliases
siat4c
Chromosome Location
Chr.8: 36264741 - 36314826 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_203337.2 NP_976082.1 8 - 9 862 61
XM_006242789.2 XP_006242851.1 9 - 10 978 61
XM_008766058.1 XP_008764280.1 8 - 9 1364 61
XM_008766059.1 XP_008764281.1 8 - 9 1353 61
XM_008766060.1 XP_008764282.1 8 - 9 1812 61
GenBank mRNA AJ626825.1 - 7 - 8 626 61
BC089057.1 - 8 - 9 862 61
FQ226042.1 - 1 - 2 107 61

View Full Product Details