Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ostn
Entrez Gene ID
360730
Gene Name
osteocrin
Gene Aliases
-
Chromosome Location
Chr.11: 76855851 - 76888140 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_207612.2 NP_997495.1 1 - 2 102 89
XM_008768801.1 XP_008767023.1 2 - 3 294 89
XM_008768803.1 XP_008767025.1 3 - 4 444 89
GenBank mRNA AY398682.1 - - 104 89
AY573934.1 - 1 - 2 102 89

View Full Product Details