The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_031315.1 | NP_112605.1 | 3 - 3 | 1244 | 158 |
GenBank mRNA | AB010428.1 | - | 3 - 3 | 1239 | 158 |
Y09334.1 | - | 3 - 3 | 1244 | 158 |