The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_145091.4 | NP_659559.2 | 2 - 2 | 1795 | 93 |
XM_006255048.2 | XP_006255110.1 | 2 - 2 | 1840 | 93 | |
GenBank mRNA | AF062741.1 | - | 2 - 2 | 1695 | 93 |
BC072485.1 | - | - | 1795 | 93 |